L-Leucine-tert-butyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride, H-Leu-OtBu·HCl, L-leucine 1,1-dimethylethyl ester hydrochloride, H-Leu-OtBu.HCl, TERT-BUTYL (2S)-2-AMINO-4-METHYLPENTANOATE HYDROCHLORIDE
H59980-03
H59980-06
CAAAH59980-03EA
49.93
CAD
CAAAH59980-03
CAAAH59980-06
L-Leucine-tert-butyl ester hydrochloride 98%
L-Leucine-tert-butyl ester hydrochloride
Purity: 98%
CAS No.: 2748-02-9
Molecular Formula: C10H21NO2.HCl
Formula Weight: 223.74
Storage Temperature: -30°C to -10°C
Beilstein Reference: 3697141
Optical Rotation: +12° (c=2 in DMF)
MDL No.: MFCD00038906
Formula:
C₁₀H₂₁NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00038906 CAS Number: 2748-02-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...