3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine, 4-[2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine

H59970-03 H59970-MD
CAAAH59970-03EA 504.02 CAD
CAAAH59970-03 CAAAH59970-MD
3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester 95%
3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester

Purity: 95% Molecular Formula: C16H23BFNO3 Formula Weight: 307.17 MDL No.: MFCD22988989

Formula: C₁₆H₂₃BFNO₃
MW: 307.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22988989
CAS Number: 873431-46-0

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR