Azetidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59935-03
H59935-06
CAAAH59935-03EA
123.08
CAD
CAAAH59935-03
CAAAH59935-06
Azetidine hydrochloride 97%
Azetidine hydrochloride
Purity: 97%
CAS No.: 36520-39-5
Molecular Formula: C3H7N.HCl
Formula Weight: 93.56
Flash Point: 152°(306°F)
Melting Point:
Formula:
C₃H₈ClN Melting Pt: >300 °C Flash Pt: 152 °C(306 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00191762 CAS Number: 36520-39-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...