(±)-2-Amino-2-(4-chlorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H59921-14 H59921-22
CAAAH59921-14EA 192.76 CAD
CAAAH59921-14 CAAAH59921-22
(±)-2-Amino-2-(4-chlorophenyl)acetic acid 98%
(±)-2-Amino-2-(4-chlorophenyl)acetic acid

CAS: 6212-33-5 MDL: MFCD00049328

Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00049328
CAS Number: 6212-33-5

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR