4-Biphenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
H59919-03
H59919-06
CAAAH59919-03EA
84.21
CAD
CAAAH59919-03
CAAAH59919-06
4-Biphenylboronic acid pinacol ester 97%
4-Biphenylboronic acid pinacol ester
CAS: 144432-80-4
MDL: MFCD07368291
Formula:
C₁₈H₂₁BO₂ Storage Temperature: Ambient |
MDL Number:
MFCD07368291 CAS Number: 144432-80-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...