4-Hydroxybenzenesulfonic acid 65% in water
Supplier: Thermo Scientific Chemicals
|
Danger
|
H59917-AK
H59917-K2
CAAAH59917-AKEA
95.43
CAD
CAAAH59917-AK
CAAAH59917-K2
4-Hydroxybenzenesulfonic acid 65% in water
4-Hydroxybenzenesulphonic acid
CAS: 98-67-9
MDL: MFCD00007506
Formula:
C₆H₆O₄S Density: 1.337 Storage Temperature: Ambient |
MDL Number:
MFCD00673723 CAS Number: 98-67-9 EINECS: 202-691-6 UN: 1803 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...