1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine 97%

Supplier: Thermo Scientific Chemicals
Warning

H59911-03 H59911-MD
CAAAH59911-03EA 390.26 CAD
CAAAH59911-03 CAAAH59911-MD
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine 97%
1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrrolidine

Purity: 97% CAS No.: 852227-90-8 Molecular Formula: C16H24BNO2 Formula Weight: 273.18 MDL No.: MFCD08060504

Formula: C₁₆H₂₄BNO₂
Storage Temperature: Ambient
MDL Number: MFCD08060504
CAS Number: 852227-90-8

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR