3-Methyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H59899-03 H59899-06
CAAAH59899-03EA 176.96 CAD
CAAAH59899-03 CAAAH59899-06
3-Methyl-1H-pyrazole-5-carboxylic acid 97%
3-Methyl-1H-pyrazole-5-carboxylic acid

Purity: 97% CAS No.: 402-61-9 Molecular Formula: C5H6N2O2 Formula Weight: 126.11 Melting Point: 236-240° MDL No.: MFCD00462235

Formula: C₅H₆N₂O₂
Melting Pt: 236…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00462235
CAS Number: 696-22-0

Order Now

Specification Test Results

Assay Minimum 97

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR