(R)-tert-Butyl-2-aminopropanoate hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-Alanine tert-butyl ester hydrochloride, tert-Butyl (R)-2-Aminopropionate Hydrochloride, (R)-2-Aminopropionic Acid tert-Butyl Ester Hydrochloride, H-D-Ala-OtBu.HCl, H-D-Ala-OtBu·HCl
H59898-03
H59898-06
CAAAH59898-03EA
141.57
CAD
CAAAH59898-03
CAAAH59898-06
(R)-tert-Butyl-2-aminopropanoate hydrochloride 98%
(R)-tert-Butyl-2-aminopropanoate hydrochloride
Purity: 98%
CAS No.: 59531-86-1
Molecular Formula: C7H15NO2.HCl
Formula Weight: 181.66
Storage Temperature: 2°C to 8°C
Beilstein Reference: 5763077
Optical Rotation: -1.4° (c=2 in ethanol)
MDL No.: MFCD00077087
Formula:
C₇H₁₅NO₂·HCl Storage Temperature: Ambient |
MDL Number:
MFCD00077087 CAS Number: 59531-86-1 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...