5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Amino-7-bromo-9-methyl-7-deazapurine
H59897-03
H59897-MD
CAAAH59897-03EA
551.42
CAD
CAAAH59897-03
CAAAH59897-MD
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine 97%
5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-4-amine
Purity: 97%
CAS No.: 1337532-51-0
Molecular Formula: C7H7BrN4
Formula Weight: 227.06
Beilstein Reference: 21981671
MDL No.: MFCD20486185
Formula:
C₇H₇BrN₄ MW: 227.065 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20486185 CAS Number: 1337532-51-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...