1-n-Butyl-3-methylimidazolium di-n-butyl phosphate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Butyl-3-methyl-1H-imidazol-3-ium dibutyl phosphate

H59893-06 H59893-18
CAAAH59893-06EA 84.21 CAD
CAAAH59893-06 CAAAH59893-18
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate 96%
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate

Purity: 96% CAS No.: 663199-28-8 Molecular Formula: C16H33N2O4P Formula Weight: 348.42 Physical Form: Liquid Refractive Index: 1.4720 Sensitivity: Moisture Sensitive Appearance: Brown MDL No.: MFCD09265043

Formula: C₁₆H₃₃N₂O₄P
MW: 348.424 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265043
CAS Number: 663199-28-8

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR