N-(tert-Butoxycarbonyl)-L-serinemethylester 95%
Supplier: Thermo Scientific Chemicals
H59889-04
H59889-09
CAAAH59889-04EA
72.05
CAD
CAAAH59889-04
CAAAH59889-09
N-(tert-Butoxycarbonyl)-L-serinemethylester 95%
N-(tert-Butoxycarbonyl)-L-serine methyl ester
Purity: 95%
CAS No.: 2766-43-0
Molecular Formula: C9H17NO5
Formula Weight: 219.23
Flash Point: 113°(235°F)
Refractive Index: 1.4525
Beilstein Reference: 3545389
Optical Rotation: -18° (c=5 in methanol)
MDL No.: MFCD00191869
Formula:
C₉H₁₇NO₅ Density: 1.082 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00191869 CAS Number: 2766-43-0 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...