N-Fmoc-β-alanine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-β-Ala-OH P, Fmoc-β-Ala-p-alkoxybenzyl alcohol, N-Fmoc-beta-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine

H59887-06 H59887-14
CAAAH59887-06EA 106.81 CAD
CAAAH59887-06 CAAAH59887-14
N-Fmoc-β-alanine 99%
N-Fmoc-β-alanine

CAS: 35737-10-1 MDL: MFCD00063328

Formula: C₁₈H₁₇NO₄
MW: 311.3 g/mol
Melting Pt: 142…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00063328
CAS Number: 35737-10-1

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR