4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-(Difluoromethoxy)-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Difluoromethoxy-2-fluorophenylboronic acid pinacol ester

H59880-03 H59880-MD
CAAAH59880-03EA 704.68 CAD
CAAAH59880-03 CAAAH59880-MD
4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester 96%
4-Difluoromethoxy-2-fluorobenzeneboronic acid pinacol ester

Purity: 96% Molecular Formula: C13H16BF3O3 Formula Weight: 288.07 MDL No.: MFCD22419269

Formula: C₁₃H₁₆BF₃O₃
MW: 288.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22419269
CAS Number: 1628442-55-6

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR