2-Amino-4,5-dimethylthiazole hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

H59878-09 H59878-18
CAAAH59878-09EA 124.5 CAD
CAAAH59878-09 CAAAH59878-18
2-Amino-4,5-dimethylthiazole hydrochloride 98%
2-Amino-4,5-dimethylthiazole hydrochloride

CAS: 71574-33-9 MDL: MFCD00012711

Formula: C₅H₉ClN₂S
Melting Pt: 273…274 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012711
CAS Number: 71574-33-9

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR