Boc-Glu-OtBu 99%

Supplier: Thermo Scientific Chemicals

H59842-03 H59842-06
CAAAH59842-03EA 111.07 CAD
CAAAH59842-03 CAAAH59842-06
Boc-Glu-OtBu 99%
Boc-Glu-OtBu

Purity: 99% CAS No.: 24277-39-2 Molecular Formula: C14H25NO6 Formula Weight: 303.36 Storage Temperature: -30°C to -10°C Beilstein Reference: 3653769 MDL No.: MFCD00038273

Formula: C14H25NO6
Storage Temperature: Refrigerator
MDL Number: MFCD00038273
CAS Number: 24277-39-2

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR