Z-Asp(OtBu)-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Cbz-Asp(OtBu)-OH, N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester

H59838-03 H59838-06
CAAAH59838-03EA 49.77 CAD
CAAAH59838-03 CAAAH59838-06
Z-Asp(OtBu)-OH 98%
Z-Asp(OtBu)-OH

Purity: 98% CAS No.: 5545-52-8 Molecular Formula: C16H21NO6 Formula Weight: 323.34 Beilstein Reference: 2569846 MDL No.: MFCD00042857

Formula: C₁₆H₂₁NO₆
MW: 323.345 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042857
CAS Number: 5545-52-8
EINECS: 226-912-0

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR