4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-(Bromomethyl)-2-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H59821-03 H59821-MD
CAAAH59821-03EA 605.14 CAD
CAAAH59821-03 CAAAH59821-MD
4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester 96%
4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester

Purity: 96% CAS No.: 1029439-49-3 Molecular Formula: C13H17BBrFO2 Formula Weight: 314.99 Melting Point: 62-64° MDL No.: MFCD18434471

Formula: C₁₃H₁₇BBrFO₂
MW: 314.989 g/mol
Melting Pt: 62…64 °C
Storage Temperature: Ambient
MDL Number: MFCD18434471
CAS Number: 1029439-49-3

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR