3-(Boc-aminomethyl)azetidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-butyl N-(azetidin-3-ylmethyl)carbamate

H59816-MD H59816-ME
CAAAH59816-MDEA 135.56 CAD
CAAAH59816-MD CAAAH59816-ME
3-(Boc-aminomethyl)azetidine hydrochloride 95%
3-(Boc-aminomethyl)azetidine hydrochloride

Purity: 95% CAS No.: 91188-15-7 Molecular Formula: C9H18N2O2.HCl Formula Weight: 222.71 MDL No.: MFCD01861760

Formula: C₉H₁₈N₂O₂
MW: 186.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861760
CAS Number: 91188-15-7

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR