2-Quinoxalinecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H59780-MC H59780-ME
CAAAH59780-MCEA 51.98 CAD
CAAAH59780-MC CAAAH59780-ME
2-Quinoxalinecarboxylic acid 97%
2-Quinoxalinecarboxylic acid

CAS: 879-65-2 MDL: MFCD00012334

Formula: C₉H₆N₂O₂
Melting Pt: 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012334
CAS Number: 879-65-2

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR