Tin(II) methanesulfonate 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

H59773-AE H59773-AK
CAAAH59773-AEEA 52.46 CAD
CAAAH59773-AE CAAAH59773-AK
Tin(II) methanesulfonate 50% (w/w) in aqueous solution
Tin(II) methanesulfonate

CAS No.: 53408-94-9 Molecular Formula: C2H6O6S2Sn Formula Weight: 308.91 Refractive Index: 1.4440 Melting Point: -27° UN No.: 3265 MDL No.: MFCD00137737

Formula: C₂H₆O₆S₂Sn
MW: 308.894 g/mol
Melting Pt: –27 °C
Density: 1.55
Storage Temperature: Ambient
MDL Number: MFCD00137737
CAS Number: 53408-94-9
EINECS: 401-640-7
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR