2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

H59738-03 H59738-MD
CAAAH59738-03EA 838.98 CAD
CAAAH59738-03 CAAAH59738-MD
2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester 96%
2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester

Purity: 96% CAS No.: 1259022-70-2 Molecular Formula: C15H20BFO4 Formula Weight: 294.13 MDL No.: MFCD18427635

Formula: C₁₅H₂₀BFO₄
MW: 294.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18427635
CAS Number: 1259022-70-2

Order Now

Specification Test Results

Assay Minimum 96


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR