FMOC-O-tert-butyl-L-tyrosine 98% (by HPLC)
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59730-06
H59730-14
CAAAH59730-06EA
105.23
CAD
CAAAH59730-06
CAAAH59730-14
FMOC-O-tert-butyl-L-tyrosine 98% (by HPLC)
FMOC-O-tert-butyl-L-tyrosine
Purity: 98%
CAS No.: 71989-38-3
Molecular Formula: C28H29NO5
Formula Weight: 459.53
Storage Temperature: 2°C to 8°C
Melting Point: 153-156°
Beilstein Reference: 4216652
Optical Rotation: -29° (c=1 in DMF)
MDL No.: MFCD00037129
Formula:
C₂₈H₂₉NO₅ Melting Pt: 153…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037129 CAS Number: 71989-38-3 EINECS: 276-262-7 |
Specification Test Results
Assay (HPLC) |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...