Monomethyl succinate 95%

Supplier: Thermo Scientific Chemicals

H59722-14 H59722-22
CAAAH59722-14EA 136.2 CAD
CAAAH59722-14 CAAAH59722-22
Monomethyl succinate 95%
Monomethyl succinate

CAS: 3878-55-5 MDL: MFCD00002788

Formula: C₅H₈O₄
Boiling Pt: 151 °C/20 mm
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00002788
CAS Number: 3878-55-5
EINECS: 223-408-2

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR