N-ɑ-Fmoc-N-Trityl-L-histidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59711-06
H59711-14
CAAAH59711-06EA
77.26
CAD
CAAAH59711-06
CAAAH59711-14
N-ɑ-Fmoc-N-Trityl-L-histidine 98%
N-alpha-Fmoc-N-Trityl-L-histidine
Purity: 98%
CAS No.: 109425-51-6
Molecular Formula: C40H33N3O4
Formula Weight: 619.71
Storage Temperature: 2°C to 8°C
Melting Point: 145°
Beilstein Reference: 5204720
Optical Rotation: +87° (c=5 in chloroform)
MDL No.: MFCD00043332
Formula:
C₄₀H₃₃N₃O₄ Melting Pt: 145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043332 CAS Number: 109425-51-6 EINECS: 439-640-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...