Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
H59696-03
H59696-06
CAAAH59696-03EA
75.37
CAD
CAAAH59696-03
CAAAH59696-06
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride 98%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride
Purity: 98%
CAS No.: 139262-23-0
Molecular Formula: C21H24N2O4.HCl
Formula Weight: 404.89
Storage Temperature: 2°C to 8°C
Beilstein Reference: 8663370
MDL No.: MFCD00190889
Formula:
C₂₁H₂₄N₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00190889 CAS Number: 139262-23-0 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...