N,N,N',N'-Tetraphenylbenzidine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TPB, N,N,N,N-Tetraphenylbenzidine, 4,4'-Bis(diphenylamino)biphenyl

H59650-03 H59650-06
CAAAH59650-03EA 66.04 CAD
CAAAH59650-03 CAAAH59650-06
N,N,N',N'-Tetraphenylbenzidine 97%
N,N,N',N'-Tetraphenylbenzidine

Purity: 97% CAS No.: 15546-43-7 Molecular Formula: C36H28N2 Formula Weight: 488.62 Physical Form: Solid Melting Point: 230-234° MDL No.: MFCD00228123

Formula: C₃₆H₂₈N₂
MW: 488.63 g/mol
Melting Pt: 230…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00228123
CAS Number: 15546-43-7
EINECS: 239-599-0

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR