3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals

H59640-03 H59640-MD
CAAAH59640-03EA 500.86 CAD
CAAAH59640-03 CAAAH59640-MD
3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester 95%
3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester

Purity: 95% Molecular Formula: C17H26BFN2O2 Formula Weight: 320.21 MDL No.: MFCD22988988

Formula: C₁₇H₂₆BFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22988988
CAS Number: 1408088-34-5

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR