(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 98%
Supplier: Thermo Scientific Chemicals
H59634-03
H59634-06
CAAAH59634-03EA
176.96
CAD
CAAAH59634-03
CAAAH59634-06
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 98%
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
Purity: 98%
CAS No.: 5399-87-1
Molecular Formula: C10H11ClN4O4
Formula Weight: 286.67
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 158-162° dec.
Beilstein Reference: 40573
MDL No.: MFCD00005738
Formula:
C₁₀H₁₁ClN₄O₄ Melting Pt: 158…162 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005738 CAS Number: 5399-87-1 EINECS: 226-438-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...