ω-Bromo-2,4-dichloroacetophenone 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromo-1-(2,4-dichlorophenyl)ethan-1-one, 2,4-Dichlorophenacyl bromide, α-Bromo-2,4-dichloroacetophenone, 2-Bromo-2',4'-dichloroacetophenone, 2-Bromo-1-(2,4-dichlorophenyl)ethanone
H59615-03
H59615-06
CAAAH59615-03EA
116.92
CAD
CAAAH59615-03
CAAAH59615-06
ω-Bromo-2,4-dichloroacetophenone 95%
ω-Bromo-2,4-dichloroacetophenone
CAS: 2631-72-3
MDL: MFCD00053005
Formula:
C₈H₅BrCl₂O Melting Pt: 25…29 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053005 CAS Number: 2631-72-3 EINECS: 220-116-7 UN: 1759 ADR: 8,III |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...