2,3-Dihydrobenzo[b]furan-5-carbaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Coumarancarboxaldehyde, 2,3-Dihydrobenzofuran-5-carbaldehyde, 2,3-Dihydrobenzo[b]furan-5-carboxaldehyde, 2,3-Dihydrobenzofuran-5-carboxaldehyde
H59602-03
H59602-06
CAAAH59602-03EA
50.24
CAD
CAAAH59602-03
CAAAH59602-06
2,3-Dihydrobenzo[b]furan-5-carbaldehyde 97%
2,3-Dihydrobenzo[b]furan-5-carbaldehyde
Purity: 97%
CAS No.: 55745-70-5
Molecular Formula: C9H8O2
Formula Weight: 148.16
Flash Point:
Formula:
C9H8O2 MW: 148.16 g/mol Boiling Pt: 88…89 °C/0.1 mm Density: 1.177 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00068058 CAS Number: 55745-70-5 EINECS: 259-788-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...