4-Bromo-3-fluorobenzyl alcohol 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59557-03
H59557-MD
CAAAH59557-03EA
372.88
CAD
CAAAH59557-03
CAAAH59557-MD
4-Bromo-3-fluorobenzyl alcohol 96%
4-Bromo-3-fluorobenzyl alcohol
Purity: 96%
CAS No.: 222978-01-0
Molecular Formula: C7H6BrFO
Formula Weight: 205.03
MDL No.: MFCD08236860
Formula:
C₇H₆BrFO Storage Temperature: Ambient |
MDL Number:
MFCD08236860 CAS Number: 222978-01-0 |
Specification Test Results
Assay Minimum | 96 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...