tert-Butyl azetidin-3-ylcarbamate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Boc-amino)azetidine
H59556-MD
H59556-ME
CAAAH59556-MDEA
132.09
CAD
CAAAH59556-MD
CAAAH59556-ME
tert-Butyl azetidin-3-ylcarbamate 98%
tert-Butyl azetidin-3-ylcarbamate
Purity: 98%
CAS No.: 91188-13-5
Molecular Formula: C8H16N2O2
Formula Weight: 172.23
Flash Point: 116°(241°F)
Boiling Point: 268°
MDL No.: MFCD01861759
Formula:
C₈H₁₆N₂O₂ MW: 172.23 g/mol Boiling Pt: 268 °C Density: 1.06 Flash Pt: 116 °C(241 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01861759 CAS Number: 91188-13-5 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...