L-Glutamic acid diethyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Diethyl L-glutamate, H-Glu(OEt)-OEt.HCl, H-Glu(OEt)-OEt·HCl

H59551-06 H59551-14
CAAAH59551-06EA 55.3 CAD
CAAAH59551-06 CAAAH59551-14
L-Glutamic acid diethyl ester hydrochloride 98%
L-Glutamic acid diethyl ester hydrochloride

Purity: 98% CAS No.: 1118-89-4 Molecular Formula: C9H17NO4.HCl Formula Weight: 239.70 Melting Point: 108-110° Beilstein Reference: 214-270- Optical Rotation: +22° (c=1 in H2O) MDL No.: MFCD00012509

Formula: C₉H₁₈ClNO₄
MW: 239.696 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00012509
CAS Number: 1118-89-4
EINECS: 214-270-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR