Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nepsilon-Benzyloxycarbonyl-Nα-Boc-L-lysine, Boc-Lys(Z)-OH, Nɑ-Boc-Nepsilon-Cbz-L-lysine, Boc-Lys(Cbz)-OH
H59537-04
H59537-09
CAAAH59537-04EA
76
CAD
CAAAH59537-04
CAAAH59537-09
Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine 95%
Nalpha-(tert-Butoxycarbonyl)-N-epsilon-carbobenzoxy-L-lysine
Purity: 95%
CAS No.: 2389-45-9
Molecular Formula: C19H28N2O6
Formula Weight: 380.44
Storage Temperature: -30°C to -10°C
Beilstein Reference: 1917222
MDL No.: MFCD00065584
Formula:
C₁₉H₂₈N₂O₆ MW: 380.44 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00065584 CAS Number: 2389-45-9 EINECS: 219-221-0 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...