Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 97% (by HPLC)

Supplier: Thermo Scientific Chemicals

Synonyms: Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine

H59536-06 H59536-14
CAAAH59536-06EA 107.28 CAD
CAAAH59536-06 CAAAH59536-14
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine 97% (by HPLC)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine

Purity: 97% CAS No.: 132388-59-1 Molecular Formula: C38H32N2O5 Formula Weight: 596.67 Storage Temperature: 2°C to 8°C Melting Point: 201-204° Beilstein Reference: 4343823 Optical Rotation: -15° (c=1 in methanol) MDL No.: MFCD00077049

Formula: C₃₈H₃₂N₂O₅
Melting Pt: 201…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00077049
CAS Number: 132388-59-1

Order Now

Specification Test Results

Assay (HPLC) Min. 96.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR