1-Methyl-3-octylimidazolium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-3-n-octylimidazolium chloride

H59534-06 H59534-18
CAAAH59534-06EA 44.4 CAD
CAAAH59534-06 CAAAH59534-18
1-Methyl-3-octylimidazolium chloride 97%
1-Methyl-3-octylimidazolium chloride

CAS: 64697-40-1 MDL: MFCD03095432

Formula: C₁₂H₂₃ClN₂
MW: 230.78 g/mol
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD03095432
CAS Number: 64697-40-1

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR