Ethyl-2-oxocyclohexanecarboxylate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59495-14
H59495-22
CAAAH59495-14EA
133.19
CAD
CAAAH59495-14
CAAAH59495-22
Ethyl-2-oxocyclohexanecarboxylate 95%
Ethyl 2-oxocyclohexanecarboxylate
CAS: 1655-07-8
MDL: MFCD00001631
Formula:
C₉H₁₄O₃ Boiling Pt: 106 °C/11 mm Density: 1.064 Flash Pt: 96 °C(205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001631 CAS Number: 1655-07-8 EINECS: 216-731-5 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...