N-Boc-S-trityl-L-cysteine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-S-trityl-L-cysteine, N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, BOC-S-trityl-L-cysteine, (R)-2-((tert-Butoxycarbonyl)amino)-3-(tritylthio)propanoic acid, Boc-Cys(Trt)-OH

H59490-06 H59490-14
CAAAH59490-06EA 152.94 CAD
CAAAH59490-06 CAAAH59490-14
N-Boc-S-trityl-L-cysteine 97%
N-Boc-S-trityl-L-cysteine

Purity: 97% CAS No.: 21947-98-8 Molecular Formula: C27H29NO4S Formula Weight: 463.59 Melting Point: 143-146° Beilstein Reference: 2028965 Optical Rotation: +28° (c=1 in ethanol) MDL No.: MFCD00038251

Formula: C₂₇H₂₉NO₄S
MW: 463.592 g/mol
Melting Pt: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00038251
CAS Number: 21947-98-8
EINECS: 244-674-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR