1-Hexyl-3-methylimidazolium iodide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-hexyl-3-methylimidazolium iodide

H59487-06 H59487-18
CAAAH59487-06EA 62.25 CAD
CAAAH59487-06 CAAAH59487-18
1-Hexyl-3-methylimidazolium iodide 98%
1-Hexyl-3-methylimidazolium iodide

CAS: 178631-05-5 MDL: MFCD12964976

Formula: C₁₀H₁₉IN₂
MW: 294.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12964976
CAS Number: 178631-05-5

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR