3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

H59485-06 H59485-14
CAAAH59485-06EA 63.99 CAD
CAAAH59485-06 CAAAH59485-14
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide 98%
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide

Purity: 98% CAS No.: 54016-70-5 Molecular Formula: C8H14BrNOS Formula Weight: 252.17 Melting Point: 81-87° Beilstein Reference: 4165775 MDL No.: MFCD00040549

Formula: C₈H₁₄BrNOS
Melting Pt: 81…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00040549
CAS Number: 54016-70-5
EINECS: 258-925-2

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR