L-Valine ethyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Val-OEt·HCl, (S)-Ethyl 2-amino-3-methylbutanoate hydrochloride

H59466-03 H59466-06
CAAAH59466-03EA 33.34 CAD
CAAAH59466-03 CAAAH59466-06
L-Valine ethyl ester hydrochloride 98%
L-Valine ethyl ester hydrochloride

Purity: 98% CAS No.: 17609-47-1 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Melting Point: 102-105° Optical Rotation: +6.7° (c=2 in H2O) MDL No.: MFCD00012511

Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00012511
CAS Number: 17609-47-1
EINECS: 241-580-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR