1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,3,5-Tri-O-benzoyl-α-D-ribofuranose, 1,3,5-Tri-O-benzoyl-a-D-ribofuranose

H59464-03 H59464-06
CAAAH59464-03EA 81.05 CAD
CAAAH59464-03 CAAAH59464-06
1,3,5-Tri-O-benzoyl-ɑ-D-ribofuranose 97%
1,3,5-Tri-O-benzoyl-alpha-D-ribofuranose

Purity: 97% CAS No.: 22224-41-5 Molecular Formula: C26H22O8 Formula Weight: 462.45 Physical Form: Powder Appearance: Pale brown Melting Point: 125-129° Beilstein Reference: 98446 Optical Rotation: +85° (c=1 in chloroform) MDL No.: MFCD00080818

Formula: C₂₆H₂₂O₈
MW: 462.45 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00080818
CAS Number: 22224-41-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR