3-Fluoroazetidine hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

H59454-MD H59454-ME
CAAAH59454-MDEA 208.56 CAD
CAAAH59454-MD CAAAH59454-ME
3-Fluoroazetidine hydrochloride 95%
3-Fluoroazetidine hydrochloride

Purity: 95% CAS No.: 617718-46-4 Molecular Formula: C3H6FN.HCl Formula Weight: 111.55 MDL No.: MFCD05663715

Formula: C3H7ClFN
Storage Temperature: Ambient
MDL Number: MFCD05663715
CAS Number: 617718-46-4

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR