Ethyl glutaryl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H59443-AA
H59443-AC
CAAAH59443-AAEA
91.01
CAD
CAAAH59443-AA
CAAAH59443-AC
Ethyl glutaryl chloride 97%
Ethyl glutaryl chloride
Purity: 97%
CAS No.: 5205-39-0
Molecular Formula: C7ClH11O3
Formula Weight: 178.61
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.139
Refractive Index: 1.4430
Appearance: Colorless
Boiling Point: 68-70°/.07mm
Beilstein Reference: 1099817
UN No.: 3265
MDL No.: MFCD00013659
Formula:
C₇H₁₁ClO₃ Boiling Pt: 68…70 °C/.07 mm Density: 1.139 Storage Temperature: Ambient |
MDL Number:
MFCD00013659 CAS Number: 5205-39-0 EINECS: 226-000-2 UN: 3265 ADR: 8,III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...