N-Fmoc-L-alanine 95%
Supplier: Thermo Scientific Chemicals
H59438-06
H59438-14
CAAAH59438-06EA
60.67
CAD
CAAAH59438-06
CAAAH59438-14
N-Fmoc-L-alanine 95%
N-Fmoc-L-alanine
Purity: 95%
CAS No.: 35661-39-3
Molecular Formula: C18H17NO4
Formula Weight: 311.34
Storage Temperature: 2°C to 8°C
Melting Point: 147-153°
Beilstein Reference: 2225975
Optical Rotation: -18° (c=1 in DMF)
MDL No.: MFCD00037139
Formula:
C₁₈H₁₇NO₄ Melting Pt: 147…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037139 CAS Number: 35661-39-3 EINECS: 252-660-6 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...