O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine 97% (by HPLC)
Supplier: Thermo Scientific Chemicals
H59422-06
H59422-14
CAAAH59422-06EA
80.74
CAD
CAAAH59422-06
CAAAH59422-14
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine 97% (by HPLC)
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine
Purity: 98%
CAS No.: 71989-33-8
Molecular Formula: C22H25NO5
Formula Weight: 383.44
Storage Temperature: 2°C to 8°C
Melting Point: 127-130°
Beilstein Reference: 3632013
Optical Rotation: +26° (c=1 in ethyl acetate)
MDL No.: MFCD00037127
Formula:
C₂₂H₂₅NO₅ Melting Pt: 127…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037127 CAS Number: 71989-33-8 EINECS: 276-260-6 |
Specification Test Results
Assay (HPLC) | Min. 97.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...