1-Ethyl-3-methylimidazolium iodide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-ethyl-3-methylimidazolium iodide

H59404-06 H59404-18
CAAAH59404-06EA 54.83 CAD
CAAAH59404-06 CAAAH59404-18
1-Ethyl-3-methylimidazolium iodide 97%
1-Ethyl-3-methylimidazolium iodide

CAS: 35935-34-3 MDL: MFCD03701101

Formula: C₆H₁₁IN₂
MW: 238.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701101
CAS Number: 35935-34-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR