2-Amino-3,6-difluorobenzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59362-03
H59362-MD
CAAAH59362-03EA
180.12
CAD
CAAAH59362-03
CAAAH59362-MD
2-Amino-3,6-difluorobenzonitrile 97%
2-Amino-3,6-difluorobenzonitrile
Purity: 97%
CAS No.: 190011-81-5
Molecular Formula: C7H4F2N2
Formula Weight: 154.12
UN No.: 3439
MDL No.: MFCD13178244
Formula:
C₇H₄F₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD13178244 CAS Number: 190011-81-5 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...