4-Fluoro-1-indanone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluoro-2,3-dihydro-1H-inden-1-one

H59356-03 H59356-06
CAAAH59356-03EA 169.06 CAD
CAAAH59356-03 CAAAH59356-06
4-Fluoro-1-indanone 97%
4-Fluoro-1-indanone

Purity: 97% CAS No.: 699-99-0 Molecular Formula: C9H7FO Formula Weight: 150.15 Melting Point: 72-76° MDL No.: MFCD00797829

Formula: C₉H₇FO
MW: 150.152 g/mol
Melting Pt: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00797829
CAS Number: 699-99-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR